Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250300.2 | NP_001237229.1 | 4 - 5 | 504 | 85 | |
GenBank mRNA | AY453393.1 | - | 4 - 5 | 512 | 85 | |
BT089208.1 | - | 4 - 5 | 506 | 85 |
Gene Symbol: LOC100781750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244335.1 | - | 4 - 5 | 511 | 85 | |
AK285254.1 | - | 4 - 5 | 505 | 85 |
Gene Symbol: LOC100782821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244265.1 | - | 4 - 5 | 505 | 85 | |
AK286217.1 | - | 4 - 5 | 505 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Glycine max Probe spans exons |