Search
Search

| Gene Symbol: STV2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251512.1 | NP_001238441.1 | 11 - 12 | 1249 | 93 | |
| GenBank mRNA | AM048817.1 | - | 11 - 12 | 1249 | 93 | |
| BT095564.1 | - | 11 - 12 | 1253 | 93 | ||
| Gene Symbol: STV2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255000.1 | NP_001241929.1 | 11 - 12 | 1216 | 93 | |
| GenBank mRNA | AM048816.1 | - | 11 - 12 | 1216 | 93 | |
Set Membership: |
|