Search
Search

BZIP44
LOC100527859| Gene Symbol: BZIP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251000.2 | NP_001237929.1 | 1 | 349 | 68 | |
| GenBank mRNA | DQ787032.1 | - | 1 | 354 | 68 | |
| Gene Symbol: LOC100527859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251629.1 | NP_001238558.1 | 1 | 345 | 68 | |
| GenBank mRNA | BT092742.1 | - | 1 | 345 | 68 | |
Set Membership: |
|