Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00153380_m1
          See other CCND2 GE Assays ›
          Gene Symbol:
          CCND2
          Gene Name:
          cyclin D2
          Gene Aliases:
          KIAK0002, MPPH3
          Chromosome Location:
          Chr.12: 4273762 - 4305353 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00153380_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCND2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001759.4 NP_001750.1 3 - 4 856 69
          GenBank mRNA AK223577.1 - 3 - 4 855 69
          BC089384.1 - 3 - 4 834 69
          BT019847.1 - 3 - 4 577 69
          D13639.1 - 3 - 4 846 69
          M90813.1 - 3 - 4 598 69
          X68452.1 - 3 - 4 732 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200006555.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000060.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human IL-10 Pathway HUMAN COLORECTAL CANCER METASTASIS HUMAN CYCLINS AND CELL CYCLE REGULATION HUMAN MOLECULAR MECHANISMS OF CANCER HUMAN BREAST CANCER REGULATION BY STATHMIN1 HUMAN PANCREATIC ADENOCARCINOMA Inventoried Human_IL-10_Pathway Human_Pancreatic_Adenocarcinoma Human_Molecular_Mechanisms_of_Cancer Human_Cyclins_and_Cell_Cycle_Regulation Human_Colorectal_Cancer_Metastasis Human_Breast_Cancer_Regulation_by_Stathmin1 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCND2

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          long-term memory
          positive regulation of cell proliferation
          adult locomotory behavior
          negative regulation of apoptotic process
          cell division
          regulation of cell cycle
          cellular response to X-ray
          positive regulation of G1/S transition of mitotic cell cycle
          positive regulation of mitotic cell cycle phase transition
          regulation of G1/S transition of mitotic cell cycle
          protein binding
          cyclin-dependent protein serine/threonine kinase regulator activity
          protein kinase binding
          protein serine/threonine kinase activator activity
          cyclin-dependent protein serine/threonine kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000541542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 ENST00000541542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000577.hg.1 ENST00000261254.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000577.hg.1 NM_001759.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000577.hg.1 TC12000060.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536537.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000541542.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 ENST00000541542.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006555.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200145072.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145072.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145072.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200145072.hg.1 ENST00000261254.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145072.hg.1 ENST00000536795.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145072.hg.1 NM_001759.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200145072.hg.1 TC1200006555.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline