Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00155479_m1
          See other CCN1 GE Assays ›
          Gene Symbol:
          CCN1
          Gene Name:
          cellular communication network factor 1
          Gene Aliases:
          CYR61, GIG1, IGFBP10
          Chromosome Location:
          Chr.1: 85580761 - 85583950 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00155479_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001554.5 NP_001545.2 2 - 3 505 88
          GenBank mRNA AF003114.1 - 1 - 2 153 88
          AF003594.1 - 2 - 3 404 88
          AF031385.1 - 2 - 3 361 88
          AK223385.1 - 2 - 3 506 88
          AK295430.1 - 2 - 3 433 88
          AK314571.1 - 2 - 3 506 88
          BC001271.1 - 2 - 3 440 88
          BC009199.2 - 2 - 3 489 88
          BC016952.1 - 2 - 3 496 88
          CR536519.1 - 2 - 3 281 88
          DA764894.1 - 2 - 3 507 88
          U62015.1 - 2 - 3 490 88
          Y11307.1 - 2 - 3 504 88
          Y12084.1 - 2 - 3 495 88
          Z98053.1 - 2 - 3 499 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008912.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000818.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCN1

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          intussusceptive angiogenesis
          atrioventricular valve morphogenesis
          apoptotic process involved in heart morphogenesis
          ventricular septum development
          apoptotic process
          chemotaxis
          cell adhesion
          signal transduction
          integrin-mediated signaling pathway
          positive regulation of biosynthetic process
          regulation of macromolecule biosynthetic process
          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          positive regulation of cell migration
          positive regulation of bone mineralization
          positive regulation of BMP signaling pathway
          positive regulation of osteoblast proliferation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of ossification
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of developmental process
          atrial septum morphogenesis
          chondroblast differentiation
          chorio-allantoic fusion
          labyrinthine layer blood vessel development
          positive regulation of cartilage development
          regulation of ERK1 and ERK2 cascade
          reactive oxygen species metabolic process
          cell-cell adhesion
          regulation of multicellular organismal development
          positive regulation of ceramide biosynthetic process
          integrin binding
          extracellular matrix structural constituent
          protein binding
          heparin binding
          growth factor binding
          extracellular matrix binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01013026.hg.1 ENST00000360431.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013026.hg.1 ENST00000451137.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013026.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013026.hg.1 ENST00000536321.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013026.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013026.hg.1 TC01000818.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013027.hg.1 ENST00000360431.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013027.hg.1 ENST00000451137.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013027.hg.1 ENST00000536321.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013027.hg.1 NM_001554.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013027.hg.1 TC01000818.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100163572.hg.1 TC0100008912.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000480413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008912.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100163573.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163573.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163573.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100163573.hg.1 ENST00000451137.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163573.hg.1 ENST00000617517.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163573.hg.1 NM_001554.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100163573.hg.1 TC0100008912.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline