Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00159576_m1
          See other NDUFA6 GE Assays ›
          Gene Symbol:
          NDUFA6
          Gene Name:
          NADH:ubiquinone oxidoreductase subunit A6
          Gene Aliases:
          B14, CI-B14, LYRM6, MC1DN33, NADHB14
          Chromosome Location:
          Chr.22: 7871 - 13117 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00159576_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NDUFA6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002490.6 NP_002481.3 1 - 2 171 90
          GenBank mRNA AF047182.1 - 1 - 2 147 90
          AK291874.1 - 1 - 2 161 90
          AK316562.1 - 1 - 2 157 90
          BC002772.1 - 1 - 2 144 90
          BG706090.1 - 1 - 2 363 90
          CR456529.1 - 1 - 2 283 90
          CR456714.1 - 1 - 2 143 90
          CR541997.1 - 1 - 2 143 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200008859.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000840.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NDUFA6

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial electron transport, NADH to ubiquinone
          aerobic respiration
          mitochondrial respiratory chain complex I assembly
          mitochondrial ATP synthesis coupled proton transport
          hydrogen ion transmembrane transport
          NADH dehydrogenase (ubiquinone) activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22014157.hg.1 ENST00000498737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22014157.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014157.hg.1 TC22000840.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014155.hg.1 ENST00000498737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22014155.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014155.hg.1 TC22000840.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22014158.hg.1 ENST00000498737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22014158.hg.1 NM_002490.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014158.hg.1 TC22000840.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008859.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164169.hg.1 TC2200008859.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164172.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164172.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164172.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164172.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164172.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164172.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164172.hg.1 TC2200008859.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164171.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164171.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164171.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164171.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164171.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164171.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164171.hg.1 TC2200008859.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164170.hg.1 TC2200008859.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000470753.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000498737.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000602404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 ENST00000617763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 NM_002490.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200164167.hg.1 TC2200008859.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0