| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015438.hg.1 |
ENST00000020945.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015438.hg.1 |
ENST00000396822.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015438.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015438.hg.1 |
TC08001199.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015437.hg.1 |
ENST00000020945.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015437.hg.1 |
ENST00000396822.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015437.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015437.hg.1 |
TC08001199.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800170553.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170553.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170553.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170553.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170553.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170553.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170553.hg.1 |
TC0800010382.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800170554.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170554.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170554.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170554.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170554.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170554.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170554.hg.1 |
TC0800010382.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010382.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170552.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170552.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170552.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170552.hg.1 |
ENST00000020945.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170552.hg.1 |
ENST00000396822.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170552.hg.1 |
NM_003068.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170552.hg.1 |
TC0800010382.hg.1 |
Clariom D TC |