Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00162091_m1
          See other SORD GE Assays ›
          Gene Symbol:
          SORD
          Gene Name:
          sorbitol dehydrogenase
          Gene Aliases:
          HEL-S-95n, HMNR8, RDH, SDH, SORD1, SORDD, XDH
          Chromosome Location:
          Chr.15: 45023195 - 45077185 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00162091_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SORD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003104.6 NP_003095.2 4 - 5 510 72
          NR_034039.2 4 - 5 595 72
          GenBank mRNA AK309672.1 - 4 - 5 595 72
          AK312444.1 - 4 - 5 574 72
          BC021085.2 - 4 - 5 551 72
          BC025295.1 - 4 - 5 499 72
          FJ224316.1 - 4 - 5 551 72
          L29008.1 - 4 - 5 558 72
          U07361.1 - 4 - 5 562 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500007107.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000350.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SORD

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          sorbitol metabolic process
          sorbitol catabolic process
          glucuronate catabolic process to xylulose 5-phosphate
          sperm motility
          fructose biosynthetic process
          L-xylitol catabolic process
          L-xylitol metabolic process
          (R,R)-butanediol dehydrogenase activity
          L-iditol 2-dehydrogenase activity
          alcohol dehydrogenase (NAD) activity
          zinc ion binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          carbohydrate binding
          identical protein binding
          D-xylulose reductase activity
          metal ion binding
          D-sorbitol dehydrogenase (acceptor) activity
          ribitol 2-dehydrogenase activity
          NAD binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15003138.hg.1 ENST00000267814.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003138.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003138.hg.1 NM_003104.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003138.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003138.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15003131.hg.1 ENST00000267814.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003131.hg.1 NM_003104.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003131.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003131.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15003132.hg.1 ENST00000267814.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003132.hg.1 NM_003104.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003132.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003132.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15003137.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003137.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15003135.hg.1 ENST00000267814.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003135.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003135.hg.1 NM_003104.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003135.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003135.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15003139.hg.1 ENST00000267814.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003139.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003139.hg.1 NM_003104.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003139.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003139.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15003133.hg.1 ENST00000267814.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003133.hg.1 NM_003104.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003133.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003133.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15003136.hg.1 ENST00000267814.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003136.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15003136.hg.1 NM_003104.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003136.hg.1 NR_034039.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15003136.hg.1 TC15000350.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500150484.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150484.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150484.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150484.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150484.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150484.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150484.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150485.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150485.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150485.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150485.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150485.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150485.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150485.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150494.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150488.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150489.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150493.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150493.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150493.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150493.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150493.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150495.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150492.hg.1 TC1500007107.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000267814.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000558580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000558789.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 ENST00000559230.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 NM_003104.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 NR_034039.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500150491.hg.1 TC1500007107.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 172779 - -
          ABI Human Genome Survey Microarray Version 2.0 203648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline