Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00165949_m1
          See other TIMP3 GE Assays ›
          Gene Symbol:
          TIMP3
          Gene Name:
          TIMP metallopeptidase inhibitor 3
          Gene Aliases:
          HSMRK222, K222, K222TA2, SFD
          Chromosome Location:
          Chr.22: 32800816 - 32863041 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00165949_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TIMP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000362.4 NP_000353.1 1 - 2 1309 59
          GenBank mRNA AK298126.1 - 2 - 3 129 59
          AK314871.1 - 1 - 2 418 59
          BC014277.2 - 1 - 2 366 3689386 59
          BT006848.1 - 1 - 2 123 59
          CR456593.1 - 1 - 2 171 59
          L15078.1 - 1 - 2 81 59
          S78453.1 - 1 - 2 399 59
          U02571.1 - 1 - 2 404 59
          U14394.1 - 1 - 2 410 59
          U67195.1 - 1 - 2 401 59
          X76227.1 - 1 - 2 193 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007150.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000248.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK298126.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Extracellular Matrix and Adhesion Molecules Human Angiogenesis HUMAN P53 MEDIATED APOPTOSIS Inventoried Human_Angiogenesis Human_p53_Mediated_Apoptosis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TIMP3

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          central nervous system development
          aging
          visual perception
          response to mechanical stimulus
          response to hormone
          response to organic substance
          tissue regeneration
          response to amino acid
          response to estrogen
          negative regulation of membrane protein ectodomain proteolysis
          response to folic acid
          negative regulation of ERK1 and ERK2 cascade
          cellular response to organic substance
          positive regulation of TRAIL-activated apoptotic signaling pathway
          negative regulation of metalloendopeptidase activity
          protease binding
          protein binding
          metalloendopeptidase inhibitor activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22004650.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004650.hg.1 ENST00000382049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004650.hg.1 ENST00000538671.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004650.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004650.hg.1 TC22000248.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004653.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004653.hg.1 ENST00000382049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004653.hg.1 ENST00000538671.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004653.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004653.hg.1 TC22000248.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22004647.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004647.hg.1 TC22000248.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22004654.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004654.hg.1 ENST00000382049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004654.hg.1 ENST00000538671.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004654.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004654.hg.1 TC22000248.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22004652.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004652.hg.1 ENST00000382049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004652.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004652.hg.1 TC22000248.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22004651.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004651.hg.1 ENST00000382049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004651.hg.1 ENST00000538671.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004651.hg.1 NM_000362.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004651.hg.1 TC22000248.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2200150343.hg.1 TC2200007150.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200150343.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150343.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150343.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150343.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150342.hg.1 TC2200007150.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200150342.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150342.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150342.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150342.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007150.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150344.hg.1 TC2200007150.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200150344.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150344.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150344.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150344.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150348.hg.1 TC2200007150.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200150348.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150348.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150348.hg.1 ENST00000266085.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200150348.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150341.hg.1 TC2200007150.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200150341.hg.1 NM_000362.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200150341.hg.1 NM_000362.4 Refseq

          Back To Top
           

          Citations (1)

          Systematic analysis of transcribed loci in ENCODE regions using RACE sequencing reveals extensive transcription in the human genome.
          Authors
          Wu JQ, Du J, Rozowsky J, Zhang Z, Urban AE, Euskirchen G, Weissman S, Gerstein M, Snyder M
          Journal
          Genome Biol - Page No R3 (Tue Jan 01 00:00:00 EST 2008)
          Volume (Year)
          9 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline