| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06017968.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06017968.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06017968.hg.1 |
TC06001548.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002917.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002917.hg.1 |
TC6_mcf_hap5000150.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003258.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003258.hg.1 |
TC6_cox_hap2000175.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001316.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001316.hg.1 |
TC6_apd_hap1000089.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002892.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002892.hg.1 |
TC6_ssto_hap7000144.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003165.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003165.hg.1 |
TC6_dbb_hap3000164.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002655.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002655.hg.1 |
TC6_mann_hap4000146.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003151.hg.1 |
NM_000434.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003151.hg.1 |
TC6_qbl_hap6000165.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600177742.hg.1 |
TC0600011463.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000375631.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000480384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000491768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011463.hg.1 |
ENST00000495807.1 |
Ensembl |