Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00169257_m1
          See other DUSP6 GE Assays ›
          Gene Symbol:
          DUSP6 POC1B-DUSP6
          Gene Name:
          dual specificity phosphatase 6
          POC1B-DUSP6 readthrough
          Gene Aliases:
          HH19, MKP3, PYST1
          -
          Chromosome Location:
          Chr.12: 89347235 - 89352501 on Build GRCh38
          Chromosome Location:
          Chr.12: 89348065 - 89526047 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00169257_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DUSP6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001946.4 NP_001937.2 2 - 3 1301 87
          GenBank mRNA AB013382.1 - 2 - 3 1190 87
          AB451265.1 - 2 - 3 839 87
          AB451393.1 - 2 - 3 839 87
          AK091753.1 - 3 - 4 1276 87
          BC003143.1 - 2 - 3 1187 87
          BC003562.1 - 2 - 3 1187 87
          BC005047.1 - 2 - 3 1263 87
          BC037236.1 - 2 - 3 1286 87
          BT006895.1 - 2 - 3 839 87
          X93920.1 - 2 - 3 1190 87
          Gene Symbol: POC1B-DUSP6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_189587.1 11 - 12 1704 87
          NR_189588.1 9 - 10 1451 87
          NR_189589.1 10 - 11 1623 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200011470.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Human MAP Kinase Pathways Inventoried Human_Map_Kinase Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DUSP6
          Click here to find siRNAs for Gene POC1B-DUSP6

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          signal transduction
          response to xenobiotic stimulus
          cell differentiation
          peptidyl-tyrosine dephosphorylation
          positive regulation of apoptotic process
          negative regulation of MAPK cascade
          response to nitrosative stress
          regulation of heart growth
          ERK1 and ERK2 cascade
          negative regulation of ERK1 and ERK2 cascade
          response to growth factor
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          protein tyrosine/serine/threonine phosphatase activity
          protein tyrosine/threonine phosphatase activity
          hydrolase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity
          MAP kinase tyrosine phosphatase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000548755.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000548755.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 ENST00000548755.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000548755.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000548755.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 ENST00000548755.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay TC1200011470.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000279488.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000308385.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000547140.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 ENST00000547291.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 NM_001946.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 NM_022652.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200188837.hg.1 TC1200011470.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline