Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00169419_m1
          See other RIPK2 GE Assays ›
          Gene Symbol:
          RIPK2
          Gene Name:
          receptor interacting serine/threonine kinase 2
          Gene Aliases:
          CARD3, CARDIAK, CCK, GIG30, RICK, RIP2
          Chromosome Location:
          Chr.8: 89757816 - 89791064 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00169419_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RIPK2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001375360.1 NP_001362289.1 7 - 8 1122 119
          NM_003821.6 NP_003812.1 8 - 9 1276 119
          XM_011517357.3 XP_011515659.1 6 - 7 649 119
          GenBank mRNA AF027706.1 - 8 - 9 1255 119
          AF064824.1 - 8 - 9 1273 119
          AF078530.1 - 8 - 9 1031 119
          AK075213.1 - 8 - 9 1249 119
          AK301448.1 - 7 - 8 1104 119
          AY358813.1 - 7 - 8 1102 119
          AY358814.1 - 8 - 9 1282 119
          AY524045.1 - 8 - 9 1118 119
          BC004553.2 - 8 - 9 1233 119
          KU178437.1 - 8 - 9 1031 119
          KU178438.1 - 7 - 8 877 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800008179.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000545.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human Protein Kinase Pathways Inventoried Human_Protein_Kinase Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RIPK2

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          positive regulation of cytokine-mediated signaling pathway
          adaptive immune response
          immune system process
          positive regulation of T-helper 1 type immune response
          apoptotic process
          inflammatory response
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          JNK cascade
          cytokine-mediated signaling pathway
          positive regulation of protein ubiquitination
          lipopolysaccharide-mediated signaling pathway
          positive regulation of chemokine production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of stress-activated MAPK cascade
          immature T cell proliferation in thymus
          positive regulation of immature T cell proliferation in thymus
          toll-like receptor 2 signaling pathway
          toll-like receptor 4 signaling pathway
          CD4-positive, alpha-beta T cell proliferation
          p38MAPK cascade
          T cell proliferation
          defense response to bacterium
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          innate immune response
          positive regulation of T-helper 1 cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          T cell receptor signaling pathway
          protein homooligomerization
          stress-activated MAPK cascade
          positive regulation of macrophage cytokine production
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing 1 signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          response to interleukin-1
          response to interleukin-12
          response to interleukin-18
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to peptidoglycan
          cellular response to muramyl dipeptide
          regulation of primary metabolic process
          protein linear polyubiquitination
          xenophagy
          cell surface toll-like receptor signaling pathway
          positive regulation of protein K63-linked ubiquitination
          positive regulation of xenophagy
          positive regulation of CD4-positive, alpha-beta T cell proliferation
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          JUN kinase kinase kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          LIM domain binding
          signaling adaptor activity
          identical protein binding
          protein homodimerization activity
          CARD domain binding
          caspase binding
          protein serine kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08007446.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08007446.hg.1 ENST00000540020.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08007446.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08007446.hg.1 TC08000545.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800156143.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800156143.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800156143.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800156143.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800156143.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800156143.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800156143.hg.1 TC0800008179.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline