Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00170899_m1
          See other GNA12 GE Assays ›
          Gene Symbol:
          GNA12
          Gene Name:
          G protein subunit alpha 12
          Gene Aliases:
          HG1M1, NNX3, RMP, gep
          Chromosome Location:
          Chr.7: 2728105 - 2844308 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00170899_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GNA12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293092.2 NP_001280021.1 1 - 2 460 69
          NM_007353.3 NP_031379.2 1 - 2 460 69
          GenBank mRNA AF493901.1 - 1 - 2 313 69
          AK295830.1 - 1 - 2 168 69
          BC087537.1 - 1 - 2 490 69
          BC111464.1 - 1 - 2 481 69
          BI553022.1 - 1 - 2 488 69
          BM557907.1 - 452 69
          L01694.1 - 1 - 2 313 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700010075.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001101.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human IL-1 Pathway HUMAN GPCR PATHWAY HUMAN THROMBIN SIGNALING HUMAN MELATONIN SIGNALING HUMAN RHOA PATHWAY 1700 Inventoried Human_IL-1_Pathway Human_Melatonin_Signaling Human_GPCR_Pathway Human_Thrombin_Signaling Human_RhoA_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GNA12

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          Rho protein signal transduction
          blood coagulation
          regulation of blood pressure
          regulation of cell shape
          response to xenobiotic stimulus
          regulation of fibroblast migration
          cell differentiation
          regulation of TOR signaling
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          intracellular signal transduction
          embryonic digit morphogenesis
          Rho-activating G protein-coupled receptor signaling pathway
          negative regulation of vascular smooth muscle cell proliferation
          negative regulation of vascular associated smooth muscle cell migration
          nucleotide binding
          G-protein coupled receptor binding
          GTPase activity
          obsolete small monomeric GTPase activity
          protein binding
          GTP binding
          guanyl nucleotide binding
          G-protein beta/gamma-subunit complex binding
          D5 dopamine receptor binding
          metal ion binding
          protein phosphatase 2A binding
          protein phosphatase activator activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000275364.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000407904.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000447791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000447791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000447791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000471281.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000471281.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000471281.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000471281.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000471281.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000471281.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000471281.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000485329.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000485329.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000485329.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000491117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 ENST00000496740.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001282441.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_001293092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010075.hg.1 NM_007353.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 109737 - -
          ABI Human Genome Survey Microarray Version 2.0 223391 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline