Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00171138_m1
          See other CXCL11 GE Assays ›
          Gene Symbol:
          CXCL11
          Gene Name:
          C-X-C motif chemokine ligand 11
          Gene Aliases:
          H174, I-TAC, IP-9, IP9, SCYB11, SCYB9B, b-R1
          Chromosome Location:
          Chr.4: 76033682 - 76036197 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00171138_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001302123.1 NP_001289052.1 1 - 2 269 64
          NM_005409.4 NP_005400.1 1 - 2 269 64
          GenBank mRNA AF002985.1 - 1 - 2 146 64
          AF030514.1 - 1 - 2 128 64
          AF352781.1 - 1 - 2 137 64
          AL703031.1 - 1 - 2 109 64
          BC005292.1 - 1 - 2 119 3934139 64
          BC012532.1 - 1 - 2 273 4096572 64
          BC110986.1 - 1 - 2 128 5178174 64
          BP289021.1 - 1 - 2 171 64
          BT006787.1 - 1 - 2 59 64
          U59286.1 - 1 - 2 81 64
          U66096.1 - 1 - 2 104 64
          Y15220.1 - 1 - 2 65 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011054.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001306.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Chemokine Panel Human Immune Response Inventoried Human_Immune Human_Chemokine Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL11

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of leukocyte chemotaxis
          chemotaxis
          inflammatory response
          immune response
          signal transduction
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          T cell chemotaxis
          positive regulation of cAMP metabolic process
          response to lipopolysaccharide
          regulation of cell proliferation
          positive regulation of cAMP-mediated signaling
          positive regulation of release of sequestered calcium ion into cytosol
          chemokine-mediated signaling pathway
          protein binding
          chemokine activity
          heparin binding
          CXCR3 chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04017896.hg.1 TC04001306.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017896.hg.1 ENST00000306621.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017896.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017896.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011054.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 ENST00000306621.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 ENST00000503860.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 NM_001302123.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 NM_005409.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400175312.hg.1 TC0400011054.hg.1 Clariom D TC

          Back To Top
           

          Citations (1)

          Novel insight into the agonistic mechanism of alefacept in vivo: differentially expressed genes may serve as biomarkers of response in psoriasis patients.
          Authors
          Haider AS, Lowes MA, Gardner H, Bandaru R, Darabi K, Chamian F, Kikuchi T, Gilleaudeau P, Whalen MS, Cardinale I, Novitskaya I, Krueger JG
          Journal
          J Immunol - Page No 7442 (Fri Jun 01 00:00:00 EDT 2007)
          Volume (Year)
          178 (2007)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline