Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00171473_m1
          See other PIM1 GE Assays ›
          Gene Symbol:
          PIM1
          Gene Name:
          Pim-1 proto-oncogene, serine/threonine kinase
          Gene Aliases:
          PIM
          Chromosome Location:
          Chr.6: 37170146 - 37175428 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00171473_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243186.1 NP_001230115.1 4 - 5 1040 77
          NM_002648.3 NP_002639.1 4 - 5 1040 77
          GenBank mRNA BC020224.1 - 4 - 5 973 4591723 77
          DQ022562.1 - 4 - 5 1109 77
          M16750.1 - 4 - 5 632 77
          M54915.1 - 4 - 5 960 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007862.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000541.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Homo sapiens Human IL-10 Pathway HUMAN IL-6 PATHWAY Inventoried Human_IL-10_Pathway Human_IL-6_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PIM1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          apoptotic process
          cell cycle
          multicellular organism development
          cell proliferation
          hyaluronan metabolic process
          negative regulation of apoptotic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          protein autophosphorylation
          vitamin D receptor signaling pathway
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          transcription factor binding
          manganese ion binding
          ribosomal small subunit binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06005931.hg.1 TC06000541.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06005931.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005931.hg.1 ENST00000468243.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005931.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06005931.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06005930.hg.1 TC06000541.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06005930.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005930.hg.1 ENST00000468243.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005930.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06005930.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600154622.hg.1 TC0600007862.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000373509.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000468243.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 ENST00000479509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_001243186.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007862.hg.1 NM_002648.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-vtt26:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0