Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172036_m1
          See other MCL1 GE Assays ›
          Gene Symbol:
          MCL1
          Gene Name:
          BCL2 family apoptosis regulator
          Gene Aliases:
          BCL2L3, EAT, MCL1-ES, MCL1L, MCL1S, Mcl-1, TM, bcl2-L-3, mcl1/EAT
          Chromosome Location:
          Chr.1: 150574551 - 150579738 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172036_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MCL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001197320.1 NP_001184249.1 2 - 3 438 124
          NM_021960.4 NP_068779.1 1 - 2 897 124
          GenBank mRNA AF118124.1 - 1 - 2 738 124
          AF118276.1 - 1 - 2 147 124
          AF118277.1 - 1 - 2 147 124
          AF118278.1 - 1 - 2 147 124
          AK294462.1 - 2 - 3 469 124
          AK297217.1 - 2 - 3 433 124
          AK300499.1 - 2 - 3 543 124
          AK304775.1 - 2 - 3 579 124
          AK312508.1 - 1 - 2 124
          AK316267.1 - 2 - 3 725 124
          BC017197.2 - 1 - 2 743 3138465 124
          BC071897.1 - 1 - 2 745 6586159 124
          BC107735.1 - 1 - 2 760 4770487 124
          BT006640.1 - 1 - 2 689 124
          FJ917536.1 - 2 - 3 124
          L08246.1 - 1 - 2 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018484.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003205.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Apoptosis Inventoried Human_Apoptosis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MCL1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          multicellular organism development
          intrinsic apoptotic signaling pathway in response to DNA damage
          apoptotic mitochondrial changes
          cellular homeostasis
          response to cytokine
          negative regulation of apoptotic process
          protein transmembrane transport
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of anoikis
          regulation of response to DNA damage stimulus
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein channel activity
          protein homodimerization activity
          protein heterodimerization activity
          BH3 domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000307940.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000307940.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000307940.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000307940.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000307940.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000307940.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000369026.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000464132.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000464132.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000464132.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000464132.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000464132.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000464132.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000464132.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 ENST00000620947.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_001197320.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_021960.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018484.hg.1 NM_182763.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline