Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172616_m1
          See other TRIM21 GE Assays ›
          Gene Symbol:
          TRIM21
          Gene Name:
          tripartite motif containing 21
          Gene Aliases:
          RNF81, RO52, Ro/SSA, SSA, SSA1
          Chromosome Location:
          Chr.11: 4384897 - 4393696 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172616_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIM21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003141.3 NP_003132.2 3 - 4 618 82
          GenBank mRNA AY742713.1 - 2 - 3 505 82
          BC010861.1 - 3 - 4 589 3900540 82
          M34551.1 - 2 - 3 543 82
          M62800.1 - 3 - 4 590 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009880.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001309.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRIM21

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          protein monoubiquitination
          cell cycle
          positive regulation of autophagy
          protein ubiquitination
          protein destabilization
          negative regulation of NF-kappaB transcription factor activity
          regulation of type I interferon production
          negative regulation of viral transcription
          response to interferon-gamma
          innate immune response
          positive regulation of cell cycle
          positive regulation of viral entry into host cell
          positive regulation of sequence-specific DNA binding transcription factor activity
          protein autoubiquitination
          interferon-gamma-mediated signaling pathway
          protein trimerization
          negative regulation of protein deubiquitination
          negative regulation of viral release from host cell
          DNA binding
          RNA binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016655.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016655.hg.1 ENST00000543625.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016655.hg.1 NM_003141.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016655.hg.1 TC11001309.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100174726.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174726.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174726.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174726.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174726.hg.1 TC1100009880.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000254436.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 ENST00000533692.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009880.hg.1 NM_003141.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline