Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172692_m1
          See other GCM1 GE Assays ›
          Gene Symbol:
          GCM1
          Gene Name:
          glial cells missing homolog 1
          Gene Aliases:
          GCMA, hGCMa
          Chromosome Location:
          Chr.6: 53125644 - 53262803 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172692_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GCM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003643.3 NP_003634.2 2 - 3 283 124
          XM_017011390.1 XP_016866879.1 6 - 7 1665 124
          GenBank mRNA AB026493.1 - 1 - 2 154 124
          AK309881.1 - 2 - 3 262 124
          BC096288.1 - 2 - 3 241 40007164 124
          D88613.1 - 2 - 3 286 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012064.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001810.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AB026493.1, BC096288.1, D88613.1, NM_003643.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Human Stem Cell Pluripotency 1700 Inventoried Human_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GCM1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          anatomical structure morphogenesis
          gliogenesis
          positive regulation of transcription from RNA polymerase II promoter
          calcium ion homeostasis
          astrocyte fate commitment
          positive regulation of syncytium formation by plasma membrane fusion
          branching involved in labyrinthine layer morphogenesis
          cell differentiation involved in embryonic placenta development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          zinc ion binding
          histone deacetylase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06021872.hg.1 TC06001810.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06021872.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06021872.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay TC0600012064.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600183074.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183074.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600183074.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183074.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600183074.hg.1 TC0600012064.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600183073.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183073.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600183073.hg.1 ENST00000259803.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183073.hg.1 NM_003643.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600183073.hg.1 TC0600012064.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 119516 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline