Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172734_m1
          See other CRY1 GE Assays ›
          Gene Symbol:
          CRY1
          Gene Name:
          cryptochrome circadian regulator 1
          Gene Aliases:
          DSPD, PHLL1
          Chromosome Location:
          Chr.12: 106991364 - 107093549 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172734_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CRY1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001413458.1 NP_001400387.1 2 - 3 853 84
          NM_001413459.1 NP_001400388.1 2 - 3 853 84
          NM_001413460.1 NP_001400389.1 2 - 3 853 84
          NM_001413461.1 NP_001400390.1 2 - 3 853 84
          NM_001413462.1 NP_001400391.1 2 - 3 853 84
          NM_001413463.1 NP_001400392.1 2 - 3 847 84
          NM_001413464.1 NP_001400393.1 3 - 4 973 84
          NM_001413465.1 NP_001400394.1 3 - 4 973 84
          NM_001413467.1 NP_001400396.1 2 - 3 853 84
          NM_001413468.1 NP_001400397.1 2 - 3 833 84
          NM_001413469.1 NP_001400398.1 3 - 4 1115 84
          NM_001413470.1 NP_001400399.1 2 - 3 853 84
          NM_004075.5 NP_004066.1 2 - 3 853 84
          NR_182152.1 2 - 3 853 84
          NR_182153.1 2 - 3 853 84
          XM_047428319.1 XP_047284275.1 2 - 3 130 84
          GenBank mRNA AK125915.1 - 4 - 5 824 84
          AK290552.1 - 2 - 3 461 84
          BC017466.1 - 2 - 3 584 84
          BC030519.1 - 2 - 3 834 84
          D83702.1 - 2 - 3 851 84
          D84657.1 - 2 - 3 265 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200011812.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001925.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001413467.1, NM_001413468.1, NM_001413469.1, NM_001413470.1, XM_047428319.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CRY1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          gluconeogenesis
          regulation of gluconeogenesis
          circadian rhythm
          response to light stimulus
          blue light signaling pathway
          response to activity
          lipid storage
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          response to insulin
          circadian regulation of gene expression
          response to glucagon
          glucose homeostasis
          regulation of circadian rhythm
          negative regulation of circadian rhythm
          signal transduction in response to DNA damage
          entrainment of circadian clock by photoperiod
          negative regulation of gluconeogenesis
          positive regulation of gluconeogenesis
          negative regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of transcription, DNA-templated
          rhythmic process
          regulation of DNA damage checkpoint
          negative regulation of glucocorticoid receptor signaling pathway
          negative regulation of glucocorticoid secretion
          nucleotide binding
          DNA binding
          double-stranded DNA binding
          deoxyribodipyrimidine photo-lyase activity
          DNA (6-4) photolyase activity
          protein binding
          photoreceptor activity
          blue light photoreceptor activity
          ligand-dependent nuclear receptor binding
          kinase binding
          protein kinase binding
          phosphatase binding
          histone deacetylase binding
          E-box binding
          FAD binding
          DNA-binding transcription factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12025096.hg.1 ENST00000008527.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12025096.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12025096.hg.1 TC12001925.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200191538.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191538.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191538.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200191538.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191538.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191538.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200191538.hg.1 TC1200011812.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 167308 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline