| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02011963.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02011963.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02011963.hg.1 |
TC02000762.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02011962.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02011962.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02011962.hg.1 |
TC02000762.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200163819.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200163819.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200163819.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200163819.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200163819.hg.1 |
TC0200009095.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200163821.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200163821.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200163821.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200163821.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200163821.hg.1 |
TC0200009095.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009095.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200163820.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200163820.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200163820.hg.1 |
ENST00000295228.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200163820.hg.1 |
NM_002193.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200163820.hg.1 |
TC0200009095.hg.1 |
Clariom D TC |