Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00173726_m1
          See other PLA2G7 GE Assays ›
          Gene Symbol:
          PLA2G7
          Gene Name:
          phospholipase A2 group VII
          Gene Aliases:
          LDL-PLA2, LP-PLA2, PAFAD, PAFAH
          Chromosome Location:
          Chr.6: 46700954 - 46735836 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00173726_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PLA2G7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001168357.1 NP_001161829.1 2 - 3 286 94
          NM_005084.3 NP_005075.3 2 - 3 377 94
          XM_005249408.4 XP_005249465.1 2 - 3 361 94
          GenBank mRNA AK290381.1 - 2 - 3 336 94
          BC038452.1 - 2 - 3 286 5216829 94
          U20157.1 - 2 - 3 269 94
          U24577.1 - 2 - 3 324 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011953.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001773.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human LDL Pathways Human Inflammation HUMAN CAMP PATHWAY HUMAN SPERM MOTILITY Inventoried Human_Inflammation Human_LDL Human_cAMP_Pathway Human_Sperm_Motility Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PLA2G7

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid catabolic process
          low-density lipoprotein particle remodeling
          lipid oxidation
          plasma lipoprotein particle oxidation
          positive regulation of inflammatory response
          positive regulation of monocyte chemotaxis
          1-alkyl-2-acetylglycerophosphocholine esterase activity
          phospholipid binding
          hydrolase activity, acting on ester bonds
          calcium-independent phospholipase A2 activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000274793.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 ENST00000537365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_001168357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011953.hg.1 NM_005084.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0