| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015897.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015897.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015897.hg.1 |
TC12001202.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015896.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015896.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015896.hg.1 |
TC12001202.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200174381.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200174381.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200174381.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200174381.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200174381.hg.1 |
TC1200009872.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200174382.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200174382.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200174382.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200174382.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200174382.hg.1 |
TC1200009872.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009872.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200174383.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200174383.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200174383.hg.1 |
ENST00000229402.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200174383.hg.1 |
NM_002258.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200174383.hg.1 |
TC1200009872.hg.1 |
Clariom D TC |