Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00175408_m1
          See other FCER1G GE Assays ›
          Gene Symbol:
          FCER1G
          Gene Name:
          Fc fragment of IgE receptor Ig
          Gene Aliases:
          FCRG
          Chromosome Location:
          Chr.1: 161215297 - 161219248 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00175408_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FCER1G
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004106.1 NP_004097.1 1 - 2 77 101
          GenBank mRNA AK307102.1 - 1 - 2 102 101
          BC033872.1 - 1 - 2 71 5219148 101
          M33195.1 - 1 - 2 77 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010341.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001382.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Human Phagocytosis of Microbes HUMAN PATHOGENESIS OF RHEUMATOID ARTHRITIS Inventoried Human_Phagocytosis_of_Microbes Human_Pathogenesis_of_Rheumatoid_Arthritis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FCER1G

          Panther Classification:

          Molecular Function -

          defense/immunity protein immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of type IIa hypersensitivity
          positive regulation of type III hypersensitivity
          positive regulation of type I hypersensitivity
          stimulatory C-type lectin receptor signaling pathway
          neutrophil activation involved in immune response
          T cell differentiation involved in immune response
          Fc receptor mediated stimulatory signaling pathway
          serotonin secretion by platelet
          phagocytosis, engulfment
          integrin-mediated signaling pathway
          blood coagulation
          regulation of platelet activation
          immunoglobulin mediated immune response
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          platelet activation
          neutrophil chemotaxis
          receptor internalization
          positive regulation of interleukin-10 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of mast cell cytokine production
          negative regulation of mast cell apoptotic process
          Fc-gamma receptor signaling pathway
          Fc-epsilon receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I
          defense response to bacterium
          positive regulation of mast cell degranulation
          innate immune response
          mast cell activation
          positive regulation of phagocytosis
          leukocyte migration
          cellular response to low-density lipoprotein particle stimulus
          protein localization to plasma membrane
          protein binding
          IgE receptor activity
          IgE binding
          IgG binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01021855.hg.1 TC01001382.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01021855.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01021855.hg.1 ENST00000367992.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01021855.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 ENST00000490414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010341.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100176075.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100176075.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100176075.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100176075.hg.1 ENST00000289902.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100176075.hg.1 ENST00000367992.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100176075.hg.1 NM_004106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100176075.hg.1 TC0100010341.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline