Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00176666_m1
          See other ITPKB GE Assays ›
          Gene Symbol:
          ITPKB HLA-F
          Gene Name:
          inositol-trisphosphate 3-kinase B
          major histocompatibility complex, class I, F
          Gene Aliases:
          IP3-3KB, IP3K, IP3K-B, IP3KB, PIG37
          CDA12, HLA-5.4, HLA-CDA12, HLAF
          Chromosome Location:
          Chr.1: 226631690 - 226739282 on Build GRCh38
          Chromosome Location:
          Chr.6: 1206571 - 1225171 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00176666_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITPKB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002221.4 NP_002212.3 4 - 5 2690 71
          GenBank mRNA AJ242780.1 - 3 - 4 2300 71
          AK289700.1 - 3 - 4 1029 71
          AY513281.1 - 4 - 5 824 71
          X57206.1 - 3 - 4 922 71
          Y18024.2 - 3 - 4 2243 71
          Gene Symbol: HLA-F
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY253269.1 - 4 - 5 824 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100017534.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003901.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITPKB
          Click here to find siRNAs for Gene HLA-F

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          myeloid cell homeostasis
          signal transduction
          cell surface receptor signaling pathway
          T cell differentiation
          inositol trisphosphate metabolic process
          inositol phosphate biosynthetic process
          negative regulation of neutrophil apoptotic process
          common myeloid progenitor cell proliferation
          positive thymic T cell selection
          thymic T cell selection
          negative regulation of myeloid cell differentiation
          positive regulation of Ras protein signal transduction
          positive regulation of alpha-beta T cell differentiation
          phosphatidylinositol phosphorylation
          cellular response to calcium ion
          nucleotide binding
          inositol hexakisphosphate kinase activity
          protein binding
          calmodulin binding
          ATP binding
          inositol-1,4,5-trisphosphate 3-kinase activity
          kinase activity
          transferase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01059470.hg.1 ENST00000272117.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01059470.hg.1 ENST00000429204.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01059470.hg.1 NM_002221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01059470.hg.1 TC01003901.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100232707.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100232707.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100232707.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100232707.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100232707.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100232707.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100232707.hg.1 TC0100017534.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000272117.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000366784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 ENST00000429204.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Affymetrix Human Clariom D Assay TC0100017534.hg.1 NM_002221.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 214309 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline