Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00178463_m1
          See other ROCK1 GE Assays ›
          Gene Symbol:
          ROCK1
          Gene Name:
          Rho associated coiled-coil containing protein kinase 1
          Gene Aliases:
          P160ROCK, ROCK-I
          Chromosome Location:
          Chr.18: 20949740 - 21111851 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00178463_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ROCK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005406.2 NP_005397.1 13 - 14 2353 121
          GenBank mRNA AB208965.1 - 5 - 6 446 121
          AK057038.1 - 13 - 14 2354 121
          AK307388.1 - 13 - 14 2168 121
          BC113114.1 - 13 - 14 1428 7961989 121
          U43195.1 - 13 - 14 1412 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800008219.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000397.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Human TGFB Panel Human NGF Pathway HUMAN PKA SIGNALING HUMAN BREAST CANCER REGULATION BY STATHMIN1 HUMAN GPCR PATHWAY HUMAN GUIDANCE CUES AND GROWTH CONE MOTILITY HUMAN THROMBIN SIGNALING HUMAN RHO FAMILY GTPASES HUMAN RHOA PATHWAY 1700 Inventoried Human_NGF_Pathway Human_TGFB Human_Guidance_Cues_and_Growth_Cone_Motility Human_GPCR_Pathway Human_PKA_Signaling Human_Thrombin_Signaling Human_Rho_Family_GTPases Human_RhoA_Pathway Human_Breast_Cancer_Regulation_by_Stathmin1 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ROCK1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          apical constriction
          protein phosphorylation
          cellular component disassembly involved in execution phase of apoptosis
          smooth muscle contraction
          leukocyte cell-cell adhesion
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          Rho protein signal transduction
          negative regulation of angiogenesis
          membrane to membrane docking
          actin cytoskeleton organization
          regulation of cell adhesion
          cortical actin cytoskeleton organization
          bleb assembly
          negative regulation of protein binding
          regulation of actin cytoskeleton organization
          negative regulation of myosin-light-chain-phosphatase activity
          negative regulation of neuron apoptotic process
          regulation of keratinocyte differentiation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          leukocyte migration
          leukocyte tethering or rolling
          myoblast migration
          regulation of stress fiber assembly
          regulation of focal adhesion assembly
          positive regulation of focal adhesion assembly
          establishment of protein localization to plasma membrane
          regulation of establishment of endothelial barrier
          negative regulation of bicellular tight junction assembly
          regulation of establishment of cell polarity
          regulation of cell motility
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          GTP-Rho binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR18004082.hg.1 ENST00000399799.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18004082.hg.1 NM_005406.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18004082.hg.1 TC18000397.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1800156144.hg.1 ENST00000399799.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1800156144.hg.1 NM_005406.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800156144.hg.1 TC1800008219.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800156144.hg.1 ENST00000399799.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1800156144.hg.1 NM_005406.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229310 - -
          ABI Human Genome Survey Microarray Version 2.0 670499 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0