Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00179099_m1
          See other MAP3K2 GE Assays ›
          Gene Symbol:
          MAP3K2
          Gene Name:
          mitogen-activated protein kinase kinase kinase 2
          Gene Aliases:
          MEKK2, MEKK2B
          Chromosome Location:
          Chr.2: 127298669 - 127343229 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00179099_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAP3K2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006609.4 NP_006600.3 11 - 12 1150 152
          GenBank mRNA AB208963.1 - 10 - 11 1056 152
          AF111105.1 - 11 - 12 1148 152
          AF239798.1 - 10 - 11 1081 152
          AK075004.1 - 5 - 6 514 152
          AK315007.1 - 12 - 13 1335 152
          BC136293.1 - 11 - 12 1119 9020280 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200014170.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002279.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Human MAP Kinase Pathways Human TNFR2 Pathway Human NGF Pathway Human Interferon Pathway Human FGF Pathway HUMAN SIGNALING IN GAP JUNCTIONS 1700 Inventoried Human_TNFR2_Pathway Human_FGF_Pathway Human_Interferon_Pathway Human_Map_Kinase Human_NGF_Pathway Human_Signaling_in_GAP_Junctions Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAP3K2

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          activation of MAPK activity
          protein phosphorylation
          activation of JUN kinase activity
          positive regulation of transcription, DNA-templated
          cellular response to mechanical stimulus
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          protein binding
          ATP binding
          protein kinase binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02036639.hg.1 TC02002279.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02036639.hg.1 ENST00000344908.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02036639.hg.1 ENST00000409947.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02036639.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409179.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409179.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409179.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200203492.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200203492.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200203492.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200203492.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200203492.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200203492.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200203492.hg.1 TC0200014170.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000344908.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409179.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409179.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409179.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 ENST00000409947.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Affymetrix Human Clariom D Assay TC0200014170.hg.1 NM_006609.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 136013 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0