Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00180054_m1
          See other RGS2 GE Assays ›
          Gene Symbol:
          RGS2
          Gene Name:
          regulator of G-protein signaling 2
          Gene Aliases:
          G0S8
          Chromosome Location:
          Chr.1: 192809039 - 192812277 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00180054_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002923.3 NP_002914.1 1 - 2 144 62
          GenBank mRNA AF493926.1 - 1 - 2 111 62
          AK313668.1 - 1 - 2 144 62
          AY971351.1 - 1 - 2 124 62
          BC007049.1 - 1 - 2 143 3681138 62
          BT007065.1 - 1 - 2 111 62
          CD700013.1 - 1 - 2 162 62
          CR457410.1 - 1 - 2 111 62
          L13463.1 - 1 - 2 143 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011022.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001624.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens HUMAN CAMP PATHWAY 1700 Inventoried Human_cAMP_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGS2

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          cell cycle
          spermatogenesis
          regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of phospholipase activity
          negative regulation of cardiac muscle hypertrophy
          positive regulation of microtubule polymerization
          negative regulation of MAP kinase activity
          positive regulation of GTPase activity
          negative regulation of cAMP-mediated signaling
          negative regulation of G-protein coupled receptor protein signaling pathway
          brown fat cell differentiation
          relaxation of cardiac muscle
          relaxation of vascular smooth muscle
          positive regulation of cardiac muscle contraction
          regulation of adrenergic receptor signaling pathway
          G-protein alpha-subunit binding
          GTPase activator activity
          protein binding
          calmodulin binding
          beta-tubulin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01025075.hg.1 TC01001624.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01025075.hg.1 ENST00000235382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025075.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025075.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01025072.hg.1 TC01001624.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01025072.hg.1 ENST00000235382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025072.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025072.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01025073.hg.1 TC01001624.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01025073.hg.1 ENST00000235382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025073.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025073.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180842.hg.1 TC0100011022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180844.hg.1 TC0100011022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180843.hg.1 TC0100011022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100180846.hg.1 TC0100011022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000235382.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000464302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000483295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 ENST00000487236.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011022.hg.1 NM_002923.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 116793 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0