Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00182260_m1
          See other PTPN1 GE Assays ›
          Gene Symbol:
          PTPN1
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 1
          Gene Aliases:
          PTP1B
          Chromosome Location:
          Chr.20: 50510321 - 50584762 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00182260_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTPN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001278618.1 NP_001265547.1 4 - 5 609 76
          NM_002827.3 NP_002818.1 5 - 6 700 76
          GenBank mRNA AK290638.1 - 5 - 6 667 76
          AK299830.1 - 4 - 5 547 76
          AK316563.1 - 5 - 6 642 76
          AU117677.1 - 5 - 6 667 76
          BC015660.2 - 5 - 6 629 4844022 76
          BC018164.1 - 5 - 6 610 4338750 76
          BT006752.1 - 5 - 6 493 76
          M31724.1 - 5 - 6 583 76
          M33689.1 - 5 - 6 565 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007695.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000403.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Human Diabetes Human Apoptotic Pathways triggered by HIV1 HUMAN PKA SIGNALING Inventoried Human_Diabetes Human_Apoptotic_Pathways_Triggered_by_HIV1 Human_PKA_Signaling Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PTPN1

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          activation of signaling protein activity involved in unfolded protein response
          activation of JUN kinase activity
          insulin receptor signaling pathway
          regulation of signal transduction
          negative regulation of signal transduction
          regulation of endocytosis
          negative regulation of vascular endothelial growth factor receptor signaling pathway
          endoplasmic reticulum unfolded protein response
          actin cytoskeleton reorganization
          regulation of intracellular protein transport
          peptidyl-tyrosine dephosphorylation
          platelet-derived growth factor receptor-beta signaling pathway
          IRE1-mediated unfolded protein response
          negative regulation of MAP kinase activity
          negative regulation of insulin receptor signaling pathway
          regulation of type I interferon-mediated signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          positive regulation of protein tyrosine kinase activity
          negative regulation of ERK1 and ERK2 cascade
          cell-cell adhesion
          regulation of hepatocyte growth factor receptor signaling pathway
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of IRE1-mediated unfolded protein response
          negative regulation of PERK-mediated unfolded protein response
          peptidyl-tyrosine dephosphorylation involved in inactivation of protein kinase activity
          positive regulation of receptor catabolic process
          protein tyrosine phosphatase activity
          insulin receptor binding
          protein binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          receptor tyrosine kinase binding
          poly(A) RNA binding
          ephrin receptor binding
          protein phosphatase 2A binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20005743.hg.1 ENST00000371621.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20005743.hg.1 ENST00000541713.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20005743.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20005743.hg.1 TC20000403.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 NM_002827.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 TC2000007695.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 ENST00000371621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 ENST00000541713.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 NM_001278618.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000153656.hg.1 NM_002827.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7zgc8:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0