Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00184092_m1
          See other DLL4 GE Assays ›
          Gene Symbol:
          DLL4
          Gene Name:
          delta like canonical Notch ligand 4
          Gene Aliases:
          AOS6, hdelta2
          Chromosome Location:
          Chr.15: 40929333 - 40939060 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00184092_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DLL4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019074.3 NP_061947.1 6 - 7 1186 78
          GenBank mRNA AB036931.1 - 6 - 7 1126 78
          AB043894.1 - 6 - 7 850 78
          AF253468.1 - 6 - 7 850 78
          AF279305.1 - 5 - 6 756 78
          AK313831.1 - 6 - 7 1181 78
          AY358894.1 - 6 - 7 885 78
          BC106950.2 - 6 - 7 986 40034887 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500006981.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000299.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DLL4

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          patterning of blood vessels
          blood vessel remodeling
          cardiac ventricle morphogenesis
          cardiac atrium morphogenesis
          ventricular trabecula myocardium morphogenesis
          pericardium morphogenesis
          signal transduction
          Notch signaling pathway
          Notch receptor processing
          visual perception
          blood circulation
          negative regulation of cell proliferation
          negative regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of gene expression
          T cell differentiation
          dorsal aorta morphogenesis
          cellular response to vascular endothelial growth factor stimulus
          cellular response to fibroblast growth factor stimulus
          negative regulation of Notch signaling pathway
          positive regulation of Notch signaling pathway
          regulation of neurogenesis
          ventral spinal cord interneuron fate commitment
          regulation of neural retina development
          Notch signaling involved in heart development
          blood vessel lumenization
          negative regulation of cell migration involved in sprouting angiogenesis
          negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          positive regulation of neural precursor cell proliferation
          Notch binding
          calcium ion binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15002132.hg.1 ENST00000249749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15002132.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15002132.hg.1 TC15000299.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500148786.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500148786.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500148786.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500148786.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500148786.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500148786.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500148786.hg.1 TC1500006981.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000249749.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000557876.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 ENST00000559440.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006981.hg.1 NM_019074.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline