| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009952.hg.1 |
ENST00000264260.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009952.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009952.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009952.hg.1 |
TC02000623.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000450855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
ENST00000497795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008675.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160655.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160655.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160655.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160655.hg.1 |
ENST00000264260.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160655.hg.1 |
ENST00000409369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160655.hg.1 |
NM_003853.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160655.hg.1 |
TC0200008675.hg.1 |
Clariom D TC |