Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00188156_m1
          See other RAB27B GE Assays ›
          Gene Symbol:
          RAB27B
          Gene Name:
          RAB27B, member RAS oncogene family
          Gene Aliases:
          C25KG
          Chromosome Location:
          Chr.18: 54717857 - 54895516 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00188156_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RAB27B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001375327.1 NP_001362256.1 4 - 5 415 78
          NM_004163.4 NP_004154.2 3 - 4 485 78
          XM_006722518.2 XP_006722581.1 5 - 6 1020 78
          XM_017025913.2 XP_016881402.1 6 - 7 1224 78
          XM_017025914.2 XP_016881403.1 5 - 6 619 78
          XM_024451232.2 XP_024307000.1 5 - 6 2870 78
          XM_047437725.1 XP_047293681.1 5 - 6 1018 78
          XM_047437726.1 XP_047293682.1 4 - 5 413 78
          GenBank mRNA AF329499.1 - 3 - 4 485 78
          AF498954.1 - 2 - 3 242 78
          AK314115.1 - 3 - 4 763 78
          BC027474.1 - 3 - 4 353 78
          U57093.2 - 3 - 4 334 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800007360.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000189.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RAB27B

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          regulation of exocytosis
          positive regulation of exocytosis
          synaptic vesicle endocytosis
          multivesicular body sorting pathway
          anterograde axonal protein transport
          nucleotide binding
          GTPase activity
          obsolete small monomeric GTPase activity
          protein binding
          GTP binding
          hydrolase activity
          GDP binding
          protein domain specific binding
          myosin V binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18002235.hg.1 ENST00000262094.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002235.hg.1 NM_004163.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18002235.hg.1 TC18000189.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586570.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000586594.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay TC1800007360.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800150483.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150483.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150483.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800150483.hg.1 ENST00000262094.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150483.hg.1 ENST00000592334.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150483.hg.1 NM_004163.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800150483.hg.1 TC1800007360.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline