| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011204.hg.1 |
ENST00000405625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011204.hg.1 |
ENST00000405625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001055.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001055.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001055.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001055.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001055.hg.1 |
TC06000099.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147969.hg.1 |
TC0600007012.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000379153.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
ENST00000612003.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001040280.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_001251901.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007012.hg.1 |
NM_004233.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011204.hg.1 |
ENST00000405625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011204.hg.1 |
ENST00000405625.2 |
Ensembl |