Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00195365_m1
          See other DNAJA2 GE Assays ›
          Gene Symbol:
          DNAJA2
          Gene Name:
          DnaJ heat shock protein family (Hsp40) member A2
          Gene Aliases:
          CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2
          Chromosome Location:
          Chr.16: 46955362 - 46973674 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00195365_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DNAJA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005880.4 NP_005871.1 3 - 4 462 76
          GenBank mRNA AJ001309.1 - 3 - 4 412 76
          AK313031.1 - 3 - 4 450 76
          BC013044.2 - 3 - 4 454 76
          BC015809.2 - 3 - 4 455 76
          Y13350.1 - 1 - 2 97 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600010179.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16001087.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Human Heat Shock Proteins 1700 Inventoried Human_Heat_Shock Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DNAJA2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          positive regulation of cell proliferation
          response to heat
          protein refolding
          ATPase activator activity
          protein binding
          ATP binding
          zinc ion binding
          Hsp70 protein binding
          heat shock protein binding
          metal ion binding
          unfolded protein binding
          chaperone binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015270.hg.1 ENST00000317089.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015270.hg.1 NM_005880.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015270.hg.1 TC16001087.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000569553.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010179.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 ENST00000317089.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 ENST00000563158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 ENST00000617000.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 NM_005880.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600178455.hg.1 TC1600010179.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 173120 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-l697b:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline