| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007239.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007239.hg.1 |
ENST00000372236.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007239.hg.1 |
ENST00000535400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007239.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007239.hg.1 |
TC06000605.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000443535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000443535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
ENST00000443535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156594.hg.1 |
TC0600008099.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000372236.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000443535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000443535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
ENST00000443535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_001291970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008099.hg.1 |
NM_006502.2 |
Refseq |