| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018506.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018506.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018506.hg.1 |
TC03001046.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000456423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000456423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018504.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018504.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018504.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018504.hg.1 |
TC03001046.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300171680.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300171680.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300171680.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300171680.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300171680.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300171680.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300171680.hg.1 |
TC0300009853.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300171682.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300171682.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300171682.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300171682.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300171682.hg.1 |
TC0300009853.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000264734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000456423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000456423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
ENST00000468220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009853.hg.1 |
NM_006580.3 |
Refseq |