Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000356003.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000359154.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000544162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000566463.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000568684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000569297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_001270453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_001270454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_007014.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_199424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000356003.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000359154.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000544162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000566463.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000568684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
ENST00000569297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_001270453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_001270454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_007014.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
NM_199424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161986.hg.1 |
TC1600008239.hg.1 |
Clariom D TC |