Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00199841_m1
          See other ADAMTS5 GE Assays ›
          Gene Symbol:
          ADAMTS5
          Gene Name:
          ADAM metallopeptidase with thrombospondin type 1 motif 5
          Gene Aliases:
          ADAM-TS 11, ADAM-TS 5, ADAM-TS5, ADAMTS-11, ADAMTS-5, ADAMTS11, ADMP-2
          Chromosome Location:
          Chr.21: 26917912 - 26967120 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00199841_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ADAMTS5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007038.4 NP_008969.2 3 - 4 2133 65
          GenBank mRNA AF141293.1 - 1 - 2 168 65
          AF142099.1 - 3 - 4 1526 65
          AK094057.1 - 3 - 4 2132 65
          BC093775.1 - 3 - 4 1488 7939620 65
          BC093777.1 - 3 - 4 1488 7939622 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007822.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000346.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ADAMTS5

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          extracellular matrix disassembly
          protein O-linked fucosylation
          defense response to bacterium
          metalloendopeptidase activity
          integrin binding
          protein binding
          heparin binding
          metallopeptidase activity
          zinc ion binding
          extracellular matrix binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21003924.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21003924.hg.1 NM_007038.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21003924.hg.1 TC21000346.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2100152740.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100152740.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100152740.hg.1 TC2100007822.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100152740.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100152740.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 ENST00000284987.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007822.hg.1 NM_007038.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0