| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005947.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005947.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005947.hg.1 |
TC13000524.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008497.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300157253.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300157253.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300157253.hg.1 |
ENST00000380680.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300157253.hg.1 |
NM_007106.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300157253.hg.1 |
TC1300008497.hg.1 |
Clariom D TC |