| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011023.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011023.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011023.hg.1 |
TC07000699.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011024.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011024.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011024.hg.1 |
TC07000699.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011021.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011021.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011021.hg.1 |
TC07000699.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011022.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011022.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011022.hg.1 |
TC07000699.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011027.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011027.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011027.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011027.hg.1 |
TC07000699.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011026.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011026.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011026.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011026.hg.1 |
TC07000699.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700162121.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162121.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162121.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162121.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162121.hg.1 |
TC0700008786.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162126.hg.1 |
TC0700008786.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700162122.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162122.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162122.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162122.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162122.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162122.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162122.hg.1 |
TC0700008786.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700162123.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162123.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162123.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162123.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162123.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162123.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162123.hg.1 |
TC0700008786.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000249356.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000465725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
ENST00000491582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008786.hg.1 |
NM_012328.2 |
Refseq |