| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019751.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019751.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019751.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019751.hg.1 |
TC10001557.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178309.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178309.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178309.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178309.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178309.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178309.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178309.hg.1 |
TC1000011521.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |