| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007920.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007920.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007920.hg.1 |
TC06000658.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008262.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157761.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157761.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157761.hg.1 |
ENST00000211314.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157761.hg.1 |
NM_014051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157761.hg.1 |
TC0600008262.hg.1 |
Clariom D TC |