| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002776.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002776.hg.1 |
TC13000234.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002778.hg.1 |
ENST00000377918.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002778.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002778.hg.1 |
TC13000234.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009074.hg.1 |
ENST00000610846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009074.hg.1 |
ENST00000610846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002781.hg.1 |
ENST00000377918.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002781.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002781.hg.1 |
TC13000234.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002775.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002775.hg.1 |
TC13000234.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002777.hg.1 |
ENST00000377918.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002777.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13002777.hg.1 |
TC13000234.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149553.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149553.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149553.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149553.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149553.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149553.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149553.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007312.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009074.hg.1 |
ENST00000610846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009074.hg.1 |
ENST00000610846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149556.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300149549.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149549.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149549.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149554.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149555.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000612954.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
ENST00000615375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149559.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300149551.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149551.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149551.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149551.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149551.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149551.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149551.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300149552.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149552.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149552.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149552.hg.1 |
ENST00000377918.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149552.hg.1 |
ENST00000484979.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149552.hg.1 |
NM_001040429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149552.hg.1 |
TC1300007312.hg.1 |
Clariom D TC |