Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00210410_m1
          See other GPX7 GE Assays ›
          Gene Symbol:
          GPX7
          Gene Name:
          glutathione peroxidase 7
          Gene Aliases:
          CL683, GPX6, GPx-7, GSHPx-7, NPGPx
          Chromosome Location:
          Chr.1: 52602371 - 52609051 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00210410_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GPX7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015696.4 NP_056511.2 2 - 3 437 59
          GenBank mRNA AF091092.1 - 1 - 2 178 59
          AF320068.2 - 2 - 3 436 59
          AK027683.1 - 2 - 3 437 59
          AY358402.1 - 2 - 3 431 59
          BC032788.1 - 2 - 3 410 5212085 59
          BI550170.1 - 2 - 3 442 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008321.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000643.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human Antioxident Mechanisms Inventoried Human_Antioxident_Mechanisms Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GPX7

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          oxidation-reduction process
          cellular oxidant detoxification
          catalase activity
          peroxidase activity
          glutathione peroxidase activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010497.hg.1 TC01000643.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01010497.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010497.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010499.hg.1 TC01000643.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01010499.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010499.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010499.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010500.hg.1 TC01000643.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01010500.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010500.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010500.hg.1 NM_015696.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010502.hg.1 TC01000643.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01010502.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010502.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159279.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159279.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159279.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159279.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159279.hg.1 TC0100008321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008321.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159285.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159285.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159285.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159285.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159285.hg.1 TC0100008321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100159286.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159286.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159286.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159286.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159286.hg.1 TC0100008321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100159282.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159282.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159282.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159282.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159282.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159282.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159282.hg.1 TC0100008321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100159283.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159283.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159283.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159283.hg.1 ENST00000361314.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159283.hg.1 ENST00000459779.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159283.hg.1 NM_015696.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100159283.hg.1 TC0100008321.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0