Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00210873_m1
          See other NSDHL GE Assays ›
          Gene Symbol:
          NSDHL
          Gene Name:
          NAD(P) dependent 3-beta-hydroxysteroid dehydrogenase NSDHL
          Gene Aliases:
          H105E3, SDR31E1, XAP104
          Chromosome Location:
          Chr.X: 152831063 - 152869729 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00210873_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NSDHL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001129765.2 NP_001123237.1 3 - 4 275 87
          NM_001441099.1 4 - 5 369 87
          NM_015922.3 NP_057006.1 2 - 3 208 87
          XM_011531178.3 XP_011529480.1 4 - 5 369 87
          XM_017029564.2 XP_016885053.1 2 - 3 281 87
          GenBank mRNA AK026549.1 - 2 - 3 194 87
          BC000245.1 - 2 - 3 225 87
          BC007816.2 - 2 - 3 275 87
          GQ900951.1 - 2 - 3 275 87
          U47105.2 - 2 - 3 304 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008759.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000743.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NSDHL

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          lipid metabolic process
          steroid biosynthetic process
          cholesterol biosynthetic process
          smoothened signaling pathway
          steroid metabolic process
          cholesterol metabolic process
          sterol biosynthetic process
          labyrinthine layer blood vessel development
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          3-beta-hydroxy-delta5-steroid dehydrogenase activity
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          3-beta-hydroxysteroid dehydrogenase (NAD+)/C4-decarboxylase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009611.hg.1 ENST00000370274.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009611.hg.1 ENST00000440023.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009611.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009611.hg.1 NM_015922.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009611.hg.1 TC0X000743.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008759.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 ENST00000370274.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 ENST00000432467.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 ENST00000440023.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 NM_001129765.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 NM_015922.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00159600.hg.1 TC0X00008759.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0