| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
NM_016062_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8001930 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8001925 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8001931 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8001927 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8001926 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8001928 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8001929 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8001924 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
218074_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11715906_a_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11715907_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11715907_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11715906_PM_a_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016374.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016374.hg.1 |
TC16001178.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016369.hg.1 |
ENST00000422424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016369.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016369.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016369.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016369.hg.1 |
TC16001178.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016373.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016373.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016373.hg.1 |
TC16001178.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010604.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181602.hg.1 |
TC1600010604.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600181603.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181603.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181603.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181603.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181603.hg.1 |
ENST00000569299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181603.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181603.hg.1 |
TC1600010604.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000422424.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000562362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000563490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000567511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
ENST00000568572.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
NM_016062.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
NR_024525.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
NR_046109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181597.hg.1 |
TC1600010604.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
218074_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
218074_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
218074_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
218074_at |
- |
- |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_016062_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw593235 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
218074_at5 |
- |
- |
| Human Genome U95 Set |
46717_at |
- |
- |
| Human Genome U95 Set |
46717_PM_at |
- |
- |
| Human Genome U133 Set |
218074_at |
- |
- |
| Human Genome U133 Set |
218074_PM_at |
- |
- |