Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00212065_m1
          See other CYB5R4 GE Assays ›
          Gene Symbol:
          CYB5R4 RIPPLY2-CYB5R4
          Gene Name:
          cytochrome b5 reductase 4
          RIPPLY2-CYB5R4 readthrough
          Gene Aliases:
          NCB5OR, cb5/cb5R, dJ676J13.1
          -
          Chromosome Location:
          Chr.6: 83859689 - 83967423 on Build GRCh38
          Chromosome Location:
          Chr.6: 83853360 - 83967423 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00212065_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYB5R4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016230.4 NP_057314.2 5 - 6 540 87
          GenBank mRNA AF169803.1 - 4 - 5 350 87
          AK313146.1 - 5 - 6 551 87
          AL832113.1 - 5 - 6 543 87
          BC025380.1 - 5 - 6 545 87
          Gene Symbol: RIPPLY2-CYB5R4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001400774.1 NP_001387703.1 6 - 7 605 87
          NR_174603.1 6 - 7 605 87
          NR_174604.1 7 - 8 667 87
          NR_174605.1 5 - 6 826 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014148.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000774.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Human CYP450 and other Oxygenases Inventoried Human CYP450 + oxygenases Human_CYP450_and_other_Oxygenases Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CYB5R4
          Click here to find siRNAs for Gene RIPPLY2-CYB5R4

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of oxygen
          superoxide metabolic process
          bicarbonate transport
          insulin secretion
          glucose homeostasis
          response to antibiotic
          cell development
          reactive oxygen species metabolic process
          cytochrome-b5 reductase activity, acting on NAD(P)H
          NAD(P)H oxidase activity
          oxidoreductase activity
          oxidoreductase activity, acting on NAD(P)H, heme protein as acceptor
          heme binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06009277.hg.1 ENST00000369681.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06009277.hg.1 NM_016230.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06009277.hg.1 TC06000774.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600199060.hg.1 ENST00000369681.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199060.hg.1 NM_016230.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600199060.hg.1 ENST00000369681.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199060.hg.1 NM_016230.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600199060.hg.1 TC0600014148.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline