| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001357.hg.1 |
ENST00000262962.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001357.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001357.hg.1 |
TC19000084.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
ENST00000596496.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006683.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146853.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146853.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146853.hg.1 |
ENST00000262962.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146853.hg.1 |
NM_018074.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146853.hg.1 |
TC1900006683.hg.1 |
Clariom D TC |