Search
Search

GNMT| Gene Symbol: GNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318865.1 | NP_001305794.1 | 1 - 2 | 222 | 106 | |
| NM_018960.5 | NP_061833.1 | 1 - 2 | 222 | 106 | ||
| NR_134899.1 | 1 - 2 | 222 | 106 | |||
| GenBank mRNA | AF101477.1 | - | 1 - 2 | 218 | 106 | |
| BC032627.1 | - | 1 - 2 | 214 | 5229272 | 106 | |
| FJ224320.1 | - | 1 - 2 | 218 | 106 | ||
| HY010451.1 | - | 1 - 2 | 223 | 106 | ||
| KU178603.1 | - | 1 - 2 | 208 | 106 | ||
| KU178604.1 | - | 1 - 2 | 208 | 106 | ||
| KU178605.1 | - | 1 - 2 | 208 | 106 | ||
| X62250.1 | - | 1 - 2 | 359 | 106 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24889139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24889129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24889132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24889145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24889140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24889134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694204 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24889146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04694218 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 49636 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 7884 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 661724 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 841790 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P7957 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P7957 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 22346 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P126087 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P126087 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 27550 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57424 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P126087 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P31769531 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P31769500 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01467577 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P31769546 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P7957 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3210368 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P7957 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04694203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04694206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04694213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04694203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04694206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04694213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24889120 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P7957 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P126087 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04694206 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04694205 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04694213 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04694203 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04694210 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04694202 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04694204 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04694203 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810513 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810509 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810514 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810510 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810511 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810506 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810512 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810507 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15810508 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P126087 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P126087 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P7957 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 2686 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 79365 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P126087 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01467577 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15810510 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694295 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694299 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15810516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694213 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694203 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694294 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694301 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15810508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15810517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694218 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694287 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694290 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15810512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694284 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04694303 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P126087 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_018960_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8119625 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8119622 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8119624 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8119623 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8119626 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8119621 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8119620 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 210328_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11731112_at | - | - |
| Affymetrix Human Genome U219 Array | 11731112_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006724.hg.1 | TC06000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006724.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006724.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006723.hg.1 | TC06000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006723.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006723.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008062.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156024.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156024.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156024.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156024.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156024.hg.1 | TC0600008062.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600156023.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156023.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156023.hg.1 | ENST00000372808.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156023.hg.1 | NM_018960.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156023.hg.1 | TC0600008062.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 210328_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 210328_at | - | - |
| Human Genome U133A 2.0 Array | 210328_PM_at | - | - |
| Human Genome U133A 2.0 Array | 210328_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_018960_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw572615 | - | - |
| Human Genome U95 Set | 35599_PM_at | - | - |
| Human Genome U95 Set | 35599_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210328_at1 | - | - |
| Human Genome U133 Set | 210328_at | - | - |
| Human Genome U133 Set | 210328_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031540 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031540 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 123375 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_24475636-S | NM_018960.4 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_24475636-S | AF101477.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_24475636-S | BC032627.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_9426-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0000060300 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0000060300 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4860370 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_9426 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1736238 | BC032627.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1736238 | NM_018960.4 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1736238 | AF101477.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1736238 | BC032627.1 | human_transcriptome |