Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00221125_m1
          See other AS3MT GE Assays ›
          Gene Symbol:
          AS3MT BORCS7-ASMT
          Gene Name:
          arsenite methyltransferase
          BORCS7-ASMT readthrough (NMD candidate)
          Gene Aliases:
          CYT19
          C10orf32-AS3MT, C10orf32-ASMT
          Chromosome Location:
          Chr.10: 102869453 - 102901899 on Build GRCh38
          Chromosome Location:
          Chr.10: 102854210 - 102901899 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00221125_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AS3MT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020682.3 NP_065733.2 6 - 7 672 73
          GenBank mRNA AF226730.1 - 6 - 7 609 73
          AK057833.1 - 6 - 7 691 73
          AK225972.1 - 6 - 7 691 73
          AK309737.1 - 6 - 7 604 73
          BC119637.1 - 6 - 7 576 73
          BC119638.2 - 6 - 7 576 73
          Gene Symbol: BORCS7-ASMT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037644.1 10 - 11 937 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000012496.hg.1, TC1000012497.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10002948.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AS3MT
          Click here to find siRNAs for Gene BORCS7-ASMT

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          toxin metabolic process
          arsonoacetate metabolic process
          methylation
          S-adenosylmethionine-dependent methyltransferase activity
          arsenite methyltransferase activity
          methylarsonite methyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10009132.hg.1 TC10002948.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10009132.hg.1 ENST00000369880.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10009132.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000186229.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000186229.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000186229.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000186229.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000186229.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000186229.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000186229.hg.1 TC1000012496.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000186253.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000186253.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000186253.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000186253.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000186253.hg.1 TC1000012497.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000369880.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 ENST00000615257.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012496.hg.1 NM_020682.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 ENST00000299353.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012497.hg.1 NR_037644.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0