| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001149.hg.1 |
NM_001003693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001149.hg.1 |
TC6_cox_hap2000075.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001092.hg.1 |
NM_001003693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001092.hg.1 |
TC6_dbb_hap3000066.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001130.hg.1 |
NM_001003693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001130.hg.1 |
TC6_qbl_hap6000066.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06003803.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06003803.hg.1 |
ENST00000503322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06003803.hg.1 |
ENST00000556581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06003803.hg.1 |
NM_001003693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06003803.hg.1 |
TC06000381.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000964.hg.1 |
NM_001003693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000964.hg.1 |
TC6_mcf_hap5000061.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001083.hg.1 |
NM_001003693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001083.hg.1 |
TC6_mann_hap4000066.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000939.hg.1 |
NM_001003693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000939.hg.1 |
TC6_ssto_hap7000062.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198135.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198135.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198135.hg.1 |
TC0600014103.hg.1 |
Clariom D TC |